ENA068331004 Enamine

6-(3-bromophenyl)-2-methylnicotinic acid

  • MDL number MFCD05881541
  • Cas Number : 791787-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15438

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15438
Availability details for
Quantity
  • Availability
  • Add to Cart