ENA068331007 Enamine

1-(pyridin-2-ylmethyl)piperazine

  • MDL number MFCD01862551
  • Cas Number : 55579-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15440
Availability details for
Quantity
  • Availability
  • Add to Cart