ENA068331011 Enamine

4,5-dichloro-2-(2-phenoxyethyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD07690607
  • Cas Number : 144172-51-0
  • Molecular Weight : 285.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15472
Availability details for
Quantity
  • Availability
  • Add to Cart