ENA068331012 Enamine

1-(propane-1-sulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD07691361
  • Cas Number : 1009517-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16035
Availability details for
Quantity
  • Availability
  • Add to Cart