ENA068331017 Enamine

2,4-dioxo-3-phenyl-1,2,3,4-tetrahydroquinazoline-7-carboxylic acid

  • MDL number MFCD04221981
  • Cas Number : 669752-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23549
Availability details for
Quantity
  • Availability
  • Add to Cart