ENA068331020 Enamine

1-(1,2-dihydroacenaphthylen-3-yl)ethan-1-one

  • MDL number MFCD00222370
  • Cas Number : 7434-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23558
Availability details for
Quantity
  • Availability
  • Add to Cart