ENA068331033 Enamine

3-[5-(4-methoxyphenyl)furan-2-yl]propanoic acid

  • MDL number MFCD02257798
  • Cas Number : 24098-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23574
Availability details for
Quantity
  • Availability
  • Add to Cart