ENA068331035 Enamine

2-nitro-4,5-dipropoxyaniline

  • MDL number MFCD02103698
  • Cas Number : 148274-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23577

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23577
Availability details for
Quantity
  • Availability
  • Add to Cart