ENA068331037 Enamine

5-(2-furyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00980805
  • Cas Number : 4447-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23580
Availability details for
Quantity
  • Availability
  • Add to Cart