ENA068331048 Enamine

[2-(2-fluorophenoxy)ethyl](methyl)amine

  • MDL number MFCD06247665
  • Cas Number : 874623-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21842
Availability details for
Quantity
  • Availability
  • Add to Cart