ENA068331069 Enamine

2-[(4-fluorophenoxy)methyl]oxirane

  • MDL number MFCD00098471
  • Cas Number : 18123-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22416
Availability details for
Quantity
  • Availability
  • Add to Cart