ENA068331085 Enamine

1-(piperazin-1-yl)cyclohexane-1-carbonitrile

  • MDL number MFCD07692336
  • Cas Number : 874623-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23158
Availability details for
Quantity
  • Availability
  • Add to Cart