ENA068331088 Enamine

N-{[5-(2-chloropropanoyl)thiophen-2-yl]methyl}acetamide

  • MDL number MFCD07691376
  • Cas Number : 874623-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15406
Availability details for
Quantity
  • Availability
  • Add to Cart