ENA068331093 Enamine

5-(propan-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD00022397
  • Cas Number : 16935-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23232
Availability details for
Quantity
  • Availability
  • Add to Cart