ENA069848385 Enamine

2-chloro-N-[(2,3-dihydro-1,4-benzodioxin-6-yl)methyl]-N-ethylacetamide

  • MDL number MFCD08239622
  • Cas Number : 878218-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22928
Availability details for
Quantity
  • Availability
  • Add to Cart