ENA069848393 Enamine

2-chloro-1-(2,4-difluorophenyl)propan-1-one

  • MDL number MFCD08444183
  • Cas Number : 138457-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23654
Availability details for
Quantity
  • Availability
  • Add to Cart