ENA069848399 Enamine

2-(4-oxo-3,4-dihydroquinazolin-3-yl)ethane-1-sulfonyl fluoride

  • MDL number MFCD08444187
  • Cas Number : 885528-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23711
Availability details for
Quantity
  • Availability
  • Add to Cart