ENA069848401 Enamine

4-(5-mercapto-1H-tetrazol-1-yl)-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one

  • MDL number MFCD07838479
  • Cas Number : 875160-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23753

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23753
Availability details for
Quantity
  • Availability
  • Add to Cart