ENA069848415 Enamine

[2-amino-1-(3-methoxyphenyl)ethyl]diethylamine

  • MDL number MFCD08444207
  • Cas Number : 885530-72-3
  • Molecular Weight : 222.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23968
Availability details for
Quantity
  • Availability
  • Add to Cart