ENA069848425 Enamine

2-bromo-1-(2-methoxy-4-methylphenyl)ethanone

  • MDL number MFCD08239631
  • Cas Number : 145964-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24109

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24109
Availability details for
Quantity
  • Availability
  • Add to Cart