ENA069848426 Enamine

1-(2-methoxy-4-methylphenyl)ethanone

  • MDL number MFCD08239634
  • Cas Number : 35633-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34...36
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24110
Availability details for
Quantity
  • Availability
  • Add to Cart