ENA069848430 Enamine

ethyl 4-(2-chloroacetyl)-1-ethyl-2,5-dimethyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD08239626
  • Cas Number : 878218-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24123
Availability details for
Quantity
  • Availability
  • Add to Cart