ENA069848431 Enamine

1-(2-chlorobenzyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08239618
  • Cas Number : 878218-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24129
Availability details for
Quantity
  • Availability
  • Add to Cart