ENA069848435 Enamine

4-(8-methyl-4-oxo-3,4-dihydroquinazolin-3-yl)butanoic acid

  • MDL number MFCD08239604
  • Cas Number : 878217-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24143
Availability details for
Quantity
  • Availability
  • Add to Cart