ENA069848436 Enamine

thieno[3,2-d]pyrimidine-4-thiol

  • MDL number MFCD08444219
  • Cas Number : 16229-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24144
Availability details for
Quantity
  • Availability
  • Add to Cart