ENA069848458 Enamine

3-(phenylamino)propanamide

  • MDL number MFCD02250783
  • Cas Number : 21017-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23559
Availability details for
Quantity
  • Availability
  • Add to Cart