ENA069848463 Enamine

5-(2-methyl-1,3-thiazol-4-yl)-2,3-dihydro-1H-indole

  • MDL number MFCD07838404
  • Cas Number : 144340-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23642
Availability details for
Quantity
  • Availability
  • Add to Cart