ENA069848473 Enamine

ethyl 3-amino-5-(1,3-dioxaindan-5-yl)thiophene-2-carboxylate

  • MDL number MFCD07847634
  • Cas Number : 877140-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23657
Availability details for
Quantity
  • Availability
  • Add to Cart