ENA069848485 Enamine

2-chloro-1-(4-methanesulfonylpiperazin-1-yl)ethan-1-one

  • MDL number MFCD07838442
  • Cas Number : 875164-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23679
Availability details for
Quantity
  • Availability
  • Add to Cart