ENA069848487 Enamine

4-bromo-1-methyl-1H-pyrrole-2-carbohydrazide

  • MDL number MFCD07838416
  • Cas Number : 875163-57-8
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23681
Availability details for
Quantity
  • Availability
  • Add to Cart