ENA069848508 Enamine

5-(furan-2-yl)-2,3-dihydro-1,3,4-oxadiazol-2-one

  • MDL number MFCD07838478
  • Cas Number : 103418-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23716
Availability details for
Quantity
  • Availability
  • Add to Cart