ENA069848512 Enamine

2-ethyl-1H-1,3-benzodiazol-1-amine

  • MDL number MFCD07838475
  • Cas Number : 90559-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23722
Availability details for
Quantity
  • Availability
  • Add to Cart