ENA069848524 Enamine

[(2E)-3-(5-amino-4-cyano-2-furyl)prop-2-enylidene]malononitrile

  • MDL number MFCD02068667
  • Cas Number : 111203-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23743

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23743
Availability details for
Quantity
  • Availability
  • Add to Cart