ENA069848525 Enamine

4-(chloromethyl)-1-phenyl-1H-pyrazole

  • MDL number MFCD02165381
  • Cas Number : 35715-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23744
Availability details for
Quantity
  • Availability
  • Add to Cart