ENA069848531 Enamine

N-benzyl-5-(chloromethyl)-1,3,4-thiadiazole-2-carboxamide

  • MDL number MFCD07838480
  • Cas Number : 875160-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23755
Availability details for
Quantity
  • Availability
  • Add to Cart