ENA069848532 Enamine

2-methanesulfonyl-1,3-benzothiazole

  • MDL number MFCD00142947
  • Cas Number : 7144-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23757
Availability details for
Quantity
  • Availability
  • Add to Cart