ENA069848540 Enamine

methyl 3-(bromomethyl)furan-2-carboxylate

  • MDL number MFCD07847868
  • Cas Number : 23268-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23772
Availability details for
Quantity
  • Availability
  • Add to Cart