ENA069848544 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]furan-2-carboxamide

  • MDL number MFCD07838487
  • Cas Number : 875160-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23778
Availability details for
Quantity
  • Availability
  • Add to Cart