ENA069848547 Enamine

5-(chloromethyl)-N-(4-chlorophenyl)-1,3,4-thiadiazole-2-carboxamide

  • MDL number MFCD07838467
  • Cas Number : 875159-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23781

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23781
Availability details for
Quantity
  • Availability
  • Add to Cart