ENA069848580 Enamine

7-methoxy-2-methyl-2,3-dihydro-1-benzofuran-5-carboxylic acid

  • MDL number MFCD07847850
  • Cas Number : 877825-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23857
Availability details for
Quantity
  • Availability
  • Add to Cart