ENA069848581 Enamine

2-phenylpyrimidin-5-amine

  • MDL number MFCD00520252
  • Cas Number : 59808-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23860

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23860
Availability details for
Quantity
  • Availability
  • Add to Cart