ENA069848584 Enamine

1-[4-methyl-7-(prop-2-en-1-yl)-2,3,7-triazatricyclo[6.4.0.0,2,6]dodeca-1(12),3,5,8,10-pentaen-5-yl]ethan-1-one

  • MDL number MFCD01455393
  • Cas Number : 332101-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23864

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-23864
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart