ENA069848588 Enamine

3-allyl-4,5-dimethoxybenzoic acid

  • MDL number MFCD07847624
  • Cas Number : 647855-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23872
Availability details for
Quantity
  • Availability
  • Add to Cart