ENA069848589 Enamine

diethylsulfamoyl chloride

  • MDL number MFCD03249508
  • Cas Number : 20588-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23873
Availability details for
Quantity
  • Availability
  • Add to Cart