ENA069848594 Enamine

3-(2-methyl-3-oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)propanoic acid

  • MDL number MFCD01834178
  • Cas Number : 352666-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23879

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23879
Availability details for
Quantity
  • Availability
  • Add to Cart