ENA069848596 Enamine

2-(1-chloroethyl)-5-(2-furyl)thieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD07843121
  • Cas Number : 877041-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23882

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23882
Availability details for
Quantity
  • Availability
  • Add to Cart