ENA069848598 Enamine

2,2,2-trifluoroethyl pyridin-2-ylmethylcarbamate

  • MDL number MFCD07847855
  • Cas Number : 877825-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23885

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23885
Availability details for
Quantity
  • Availability
  • Add to Cart