ENA069848599 Enamine

5-(methylthio)-2-nitrobenzoic acid

  • MDL number MFCD00130056
  • Cas Number : 68701-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23886
Availability details for
Quantity
  • Availability
  • Add to Cart