ENA069848600 Enamine

octahydroimidazo[1,5-a]pyridine-1,3-dione

  • MDL number MFCD07843119
  • Cas Number : 4705-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23887
Availability details for
Quantity
  • Availability
  • Add to Cart