ENA069848613 Enamine

(4-hydroxyphenyl)(4-methylpiperazin-1-yl)acetonitrile

  • MDL number MFCD07847854
  • Cas Number : 877825-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23902
Availability details for
Quantity
  • Availability
  • Add to Cart